Anti-GPAM Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Lieferant: Bioss
BS-5063R-A350
BOSSBS-5063R-A350EA 893 EUR
BOSSBS-5063R-A350
Anti-GPAM Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Esterifies acyl-group from acyl-ACP to the sn-1 position of glycerol-3-phosphate, an essential step in glycerolipid biosynthesis.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: GPAM
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat