Anti-GPAM Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-5063R-A647
BOSSBS-5063R-A647EA 893 EUR
BOSSBS-5063R-A647
Anti-GPAM Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
Esterifies acyl-group from acyl-ACP to the sn-1 position of glycerol-3-phosphate, an essential step in glycerolipid biosynthesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GPAM
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR