Anti-IDI1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss
BS-5073R-FITC
BOSSBS-5073R-FITCEA 893 EUR
BOSSBS-5073R-FITC
Anti-IDI1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
IDI1 is a peroxisomally localized enzyme that catalyzes the interconversion of isopentenyl diphosphate (IPP) to its highly electrophilic isomer, dimethylallyl diphosphate (DMAPP). These are substrates for the reactions that ultimately result in the synthesis of cholesterol. There is reduction in IPP isomerase activity in peroxisomal deficiency diseases such as Zellweger syndrome and neonatal adrenoleukodystrophy.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IDI1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR