Anti-Ceramide synthase 2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-5077R-A750
BOSSBS-5077R-A750EA 889 EUR
BOSSBS-5077R-A750
Anti-Ceramide synthase 2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
LASS2 (LAG1 homolog, ceramide synthase 2) has sequence similarity to yeast longevity assurance gene 1. It is thought to be involved in sphingolipid synthesis. Expression of LASS2 is transiently increased during the period of active myelination and is specifically localised to white matter tracts of the brain, including the Schwann cells of sciatic nerves suggesting that LASS2 is important for the synthesis of dihydroceramide used for synthesis of myelin sphingolipids. Expression of LASS2 in hepatocellular carcinoma cell lines suppresses the growth of cancer cells.
Type: Primary
Antigen: Ceramide synthase 2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 201-300/380
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat