Anti-GPRC5C Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Lieferant: Bioss
BS-5082R-A555
BOSSBS-5082R-A555EA 893 EUR
BOSSBS-5082R-A555
Anti-GPRC5C Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
Lass5, or LAG1 longevity assurance homolog 5, is thought to be either a bona fide (dihydro)ceramide synthase or a modulator of its activity. When overexpressed in cells is involved in the production of sphingolipids containing mainly one fatty acid donnor ceramide) in a fumonisin B1-independent manner.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LASS5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR