Anti-LASS5 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-5082R-A750
BOSSBS-5082R-A750EA 890 EUR
BOSSBS-5082R-A750
Anti-LASS5 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Lass5, or LAG1 longevity assurance homolog 5, is thought to be either a bona fide (dihydro)ceramide synthase or a modulator of its activity. When overexpressed in cells is involved in the production of sphingolipids containing mainly one fatty acid donnor ceramide) in a fumonisin B1-independent manner.

Type: Primary
Antigen: LASS5
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/392
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR