Anti-C7ORF65 Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-5087R
BOSSBS-5087REA 1090 EUR
BOSSBS-5087R
Anti-C7ORF65 Rabbit Polyclonal Antibody
Antikörper
Endocannaboids such as arachidonoyl ethanolamide and 2-arachidonoyl glycerol function as short range modulators of cell and synaptic activity. Monoacylglycerol lipase hydrolyzes 2-arachidonoyl glycerol which terminates it's biological actions. Monoacylglycerol lipase also works consecutively with hormone sensitive lipase to mobilize fatty acids from triglyceride stores of adipocytes. Monoacylglycerol lipase is expressed in kidney, spleen, heart, liver, testis, stomach, brain and lung tissue, and adrenal gland. It is most abundantly expressed in skeletal muscle and adipose tissue.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: C7ORF65
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR