Anti-OXCT1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-5089R-A750
BOSSBS-5089R-A750EA 890 EUR
BOSSBS-5089R-A750
Anti-OXCT1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Key enzyme for ketone body catabolism. Transfers the CoA moiety from succinate to acetoacetate. Formation of the enzyme-CoA intermediate proceeds via an unstable anhydride species formed between the carboxylate groups of the enzyme and substrate.
Type: Primary
Antigen: OXCT1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 211-310/520
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen