Anti-BAG2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-5134R-A647
BOSSBS-5134R-A647EA 893 EUR
BOSSBS-5134R-A647
Anti-BAG2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
BAG proteins compete with Hip for binding to the Hsc70/Hsp70 ATPase domain and promote substrate release. All the BAG proteins have an approximately 45-amino acid BAG domain near the C terminus but differ markedly in their N-terminal regions. The predicted BAG2 protein contains 211 amino acids. The BAG domains of BAG1, BAG2, and BAG3 interact specifically with the Hsc70 ATPase domain in vitro and in mammalian cells. All 3 proteins bind with high affinity to the ATPase domain of Hsc70 and inhibit its chaperone activity in a Hip-repressible manner. [provided by RefSeq].

RecommendedMore Product Information
Bestellen

TECHNISCHE DATEN

Antigen-Symbol BAG2
Antigen-Name BCL2-Associated Athanogene 2
Environmentally Preferable
Antigen-Synonyme RP3-496N17.2|dJ417I1.2 (BAG-family molecular chaperone regulator 2)|KIAA0576|dJ417I1.2|BAG-family molecular chaperone regulator-2|BCL2-associated athanogene 2|BAG2|BAG-2|MGC149462
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation Alexa Fluor® 647
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Gen-ID 9532
Isotyp IgG
ImmunoChemie Yes
Immunogen 160-210/211
Form liquid
Reinigung Purified by Protein A
Konzentration 1 μg/μl
Kreuzadsorption No
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Lagertemperatur Store at 4°C for 12 months
Transporttemperatur 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR