Anti-LZTFL1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Lieferant: Bioss
BS-5138R-A350
BOSSBS-5138R-A350EA 893 EUR
BOSSBS-5138R-A350
Anti-LZTFL1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Regulates ciliary localization of the BBSome complex. Together with the BBSome complex, controls SMO ciliary trafficking and contributes to the sonic hedgehog (SHH) pathway regulation. May play a role in neurite outgrowth. May have tumor suppressor function.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: LZTFL1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat