Anti-LZTFL1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-5138R-A680
BOSSBS-5138R-A680EA 890 EUR
BOSSBS-5138R-A680
Anti-LZTFL1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Regulates ciliary localization of the BBSome complex. Together with the BBSome complex, controls SMO ciliary trafficking and contributes to the sonic hedgehog (SHH) pathway regulation. May play a role in neurite outgrowth. May have tumor suppressor function.
Type: Primary
Antigen: LZTFL1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/299
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat