Anti-LZTFL1 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-5138R-CY5.5
BOSSBS-5138R-CY5.5EA 900 EUR
BOSSBS-5138R-CY5.5
Anti-LZTFL1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Regulates ciliary localization of the BBSome complex. Together with the BBSome complex, controls SMO ciliary trafficking and contributes to the sonic hedgehog (SHH) pathway regulation. May play a role in neurite outgrowth. May have tumor suppressor function.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LZTFL1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR