Anti-MATR3 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-5141R-CY5.5
BOSSBS-5141R-CY5.5EA 900 EUR
BOSSBS-5141R-CY5.5
Anti-MATR3 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Matrin 3 (MATR 3) is an internal nuclear matrix protein that may play a role in transcription or may interact with other nuclear matrix proteins to form the internal fibrogranular network. In association with the SFPQ-NONO heteromer MATR 3 may play a role in nuclear retention of defective RNAs. MATR3 forms part of complex consisting of SFPQ, NONO and MATR3.The protein contains 1 matrin type zinc finger and 2 RRM (RNA recognition motif) domains. Two transcript variants encoding the same protein have been identified for this gene.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MATR3
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR