Anti-CAPZA1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-5161R-A488
BOSSBS-5161R-A488EA 893 EUR
BOSSBS-5161R-A488
Anti-CAPZA1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
F-actin-capping proteins bind in a Ca(2+)-independent manner to the fast growing ends of actin filaments (barbed end) thereby blocking the exchange of subunits at these ends. Unlike other capping proteins (such as gelsolin and severin), these proteins do not sever actin filaments.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CAPZA1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR