Anti-CAPZA1 Rabbit Polyclonal Antibody (Cy5.5®)
Lieferant: Bioss
BS-5161R-CY5.5
BOSSBS-5161R-CY5.5EA 893 EUR
BOSSBS-5161R-CY5.5
Anti-CAPZA1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
F-actin-capping proteins bind in a Ca(2+)-independent manner to the fast growing ends of actin filaments (barbed end) thereby blocking the exchange of subunits at these ends. Unlike other capping proteins (such as gelsolin and severin), these proteins do not sever actin filaments.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CAPZA1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human, Mouse, Rat