Anti-ATF2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-5169R-HRP
BOSSBS-5169R-HRPEA 930 EUR
BOSSBS-5169R-HRP
Anti-ATF2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
ATF2 is a member of the ATF/CREB family of basic region leucine zipper DNA binding proteins that regulates transcription by binding to a consensus cAMP response element (CRE) in the promoter of various viral and cellular genes. Many of these genes are important in cell growth and differentiation, and in stress and immune responses. ATF2 is a nuclear protein that binds DNA as a dimer and can form dimers with members of the ATF/CREB and Jun/Fos families. It is a stronger activator as a heterodimer with cJun than as a homodimer. Several isoforms of ATF2 arise by differential splicing. TheMore Product Information
Bestellen

TECHNISCHE DATEN

Antigen-Symbol ATF2
Environmentally Preferable
Antigen-Name Activating Transcription Factor 2
Antigen-Synonyme Cyclic AMP-dependent transcription factor ATF-2|HB16|cAMP-dependent transcription factor ATF-2|CREBP1|Histone acetyltransferase ATF2|ATF-2|CREB2|cAMP-responsive element-binding protein 2|CREB-2|cAMP response element-binding protein CRE-BP1|Activating transcription factor 2|Cyclic AMP-responsive element-binding protein 2
Antikörper-Typ Primär
Klonalität Polyklonal
Konjugation HRP (Horseradish Peroxidase)
Reaktivität Human,Rat,Mouse
Wirt Rabbit
Gen-ID 11909
Isotyp IgG
Western blot Yes
ImmunoChemie Yes
Modification phospho-Ser322
Form liquid
Reinigung Purified by Protein A
Konzentration 1 μg/μl
Kreuzadsorption No
Lagerungspuffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
Lagertemperatur Store at 4°C for 12 months
Transporttemperatur 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR