Anti-PTPRA Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-5175R-A350
BOSSBS-5175R-A350EA 922 EUR
BOSSBS-5175R-A350
Anti-PTPRA Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
Vital cellular functions such as cell proliferation and signal transduction are regulated in part by the balance between the activities of protein tyrosine kinases (PTK) and protein tyrosine phosphatases (PTPs). The protein contains a 121 residue extracellular domain, a single transmembrane segment, and 2 tandem intracytoplasmic catalytic domains. The PTP alpha protein is ubiquitously expressed and may play a fundamental role in the physiology of all cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PTPRA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR