Anti-PTPRA Ser180 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-5175R-A680
BOSSBS-5175R-A680EA 921 EUR
BOSSBS-5175R-A680
Anti-PTPRA Ser180 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Vital cellular functions such as cell proliferation and signal transduction are regulated in part by the balance between the activities of protein tyrosine kinases (PTK) and protein tyrosine phosphatases (PTPs). The protein contains a 121 residue extracellular domain, a single transmembrane segment, and 2 tandem intracytoplasmic catalytic domains. The PTP alpha protein is ubiquitously expressed and may play a fundamental role in the physiology of all cells.
Type: Primary
Antigen: PTPRA Ser180
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen