Anti-CDK2 Thr14 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Lieferant: Bioss
BS-5277R-A680
BOSSBS-5277R-A680EA 893 EUR
BOSSBS-5277R-A680
Anti-CDK2 Thr14 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
The protein encoded by this gene is a member of the Ser/Thr protein kinase family. This protein kinase is highly similar to the gene products of S. cerevisiae cdc28, and S. pombe cdc2. It is a catalytic subunit of the cyclin-dependent protein kinase complex, whose activity is restricted to the G1-S phase, and essential for cell cycle G1/S phase transition. This protein associates with and regulated by the regulatory subunits of the complex including cyclin A or E, CDK inhibitor p21Cip1 (CDKN1A) and p27Kip1 (CDKN1B). Its activity is also regulated by its protein phosphorylation. Two alternatively spliced variants and multiple transcription initiation sites of this gene have been reported. cdk2 is a cell cycle protein closely related to Cdc2 (cdk1) that has proved useful as a marker of proliferation. cdk2 binds cyclin type A and E proteins and controls progression into S-phase.

Type: Primary
Antigen: CDK2 Thr14
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR