Anti-G3BP1 Rabbit Polyclonal Antibody (Cy3®)
Lieferant: Bioss
BS-5376R-CY3
BOSSBS-5376R-CY3EA 902 EUR
BOSSBS-5376R-CY3
Anti-G3BP1 Rabbit Polyclonal Antibody (Cy3®)
Antikörper
May be a regulated effector of stress granule assembly. Phosphorylation-dependent sequence-specific endoribonuclease in vitro. Cleaves exclusively between cytosine and adenine and cleaves MYC mRNA preferentially at the 3'-UTR. ATP- and magnesium-dependent helicase. Unwinds preferentially partial DNA and RNA duplexes having a 17 bp annealed portion and either a hanging 3' tail or hanging tails at both 5'- and 3'-ends. Unwinds DNA/DNA, RNA/DNA, and RNA/RNA substrates with comparable efficiency. Acts unidirectionally by moving in the 5' to 3' direction along the bound single-stranded DNA.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: G3BP1Ser232
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Pig