Anti-G3BP1 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-5376R-CY5.5
BOSSBS-5376R-CY5.5EA 895 EUR
BOSSBS-5376R-CY5.5
Anti-G3BP1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
May be a regulated effector of stress granule assembly. Phosphorylation-dependent sequence-specific endoribonuclease in vitro. Cleaves exclusively between cytosine and adenine and cleaves MYC mRNA preferentially at the 3'-UTR. ATP- and magnesium-dependent helicase. Unwinds preferentially partial DNA and RNA duplexes having a 17 bp annealed portion and either a hanging 3' tail or hanging tails at both 5'- and 3'-ends. Unwinds DNA/DNA, RNA/DNA, and RNA/RNA substrates with comparable efficiency. Acts unidirectionally by moving in the 5' to 3' direction along the bound single-stranded DNA.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: G3BP1Ser232
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR