Anti-NFKBIA Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-5515R-CY5
BOSSBS-5515R-CY5EA 929 EUR
BOSSBS-5515R-CY5
Anti-NFKBIA Rabbit Polyclonal Antibody (Cy5®)
Antikörper
Inhibits the activity of dimeric NF-kappa-B/REL complexes by trapping REL dimers in the cytoplasm through masking of their nuclear localization signals. On cellular stimulation by immune and proinflammatory responses, becomes phosphorylated promoting ubiquitination and degradation, enabling the dimeric RELA to translocate to the nucleus and activate transcription.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: NFKBIA
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat