Anti-NFKBIA Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-5515R-CY5.5
BOSSBS-5515R-CY5.5EA 929 EUR
BOSSBS-5515R-CY5.5
Anti-NFKBIA Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Inhibits the activity of dimeric NF-kappa-B/REL complexes by trapping REL dimers in the cytoplasm through masking of their nuclear localization signals. On cellular stimulation by immune and proinflammatory responses, becomes phosphorylated promoting ubiquitination and degradation, enabling the dimeric RELA to translocate to the nucleus and activate transcription.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NFKBIA
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR