Anti-PIK3C3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-5581R-A488
BOSSBS-5581R-A488EA 895 EUR
BOSSBS-5581R-A488
Anti-PIK3C3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Catalytic subunit of the PI3K complex that mediates formation of phosphatidylinositol 3-phosphate which plays a key role in initiation and maturation of autophagosomes. Involved in the transport of lysosomal enzyme precursors to lysosomes. Required for the abcission step in cytokinesis. Required for transport from early to late endosomes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PIK3C3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR