Anti-RASGRF1 Ser929 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-5683R-A750
BOSSBS-5683R-A750EA 921 EUR
BOSSBS-5683R-A750
Anti-RASGRF1 Ser929 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
The protein encoded by this gene is a guanine nucleotide exchange factor (GEF) similar to the Saccharomyces cerevisiae CDC25 gene product. Functional analysis has demonstrated that this protein stimulates the dissociation of GDP from RAS protein. The studies of the similar gene in mouse suggested that the Ras-GEF activity of this protein in brain can be activated by Ca2+ influx, muscarinic receptors, and G protein beta-gamma subunit. Mouse studies also indicated that the Ras-GEF signaling pathway mediated by this protein may be important for long-term memory. Alternatively spliced transcript variants encoding distinct isoforms have been reported.

Type: Primary
Antigen: RASGRF1 Ser929
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR