Anti-SCNN1B Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-5701R-CY7
BOSSBS-5701R-CY7EA 930 EUR
BOSSBS-5701R-CY7
Anti-SCNN1B Rabbit Polyclonal Antibody (Cy7®)
Antikörper
Sodium permeable non-voltage-sensitive ion channel inhibited by the diuretic amiloride. Mediates the electrodiffusion of the luminal sodium (and water, which follows osmotically) through the apical membrane of epithelial cells. Plays an essential role in electrolyte and blood pressure homeostasis, but also in airway surface liquid homeostasis, which is important for proper clearance of mucus. Controls the reabsorption of sodium in kidney, colon, lung and sweat glands. Also plays a role in taste perception.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SCNN1B
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR