Anti-SCNN1B Ser633 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-5702R-A750
BOSSBS-5702R-A750EA 917 EUR
BOSSBS-5702R-A750
Anti-SCNN1B Ser633 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Sodium permeable non-voltage-sensitive ion channel inhibited by the diuretic amiloride. Mediates the electrodiffusion of the luminal sodium (and water, which follows osmotically) through the apical membrane of epithelial cells. Plays an essential role in electrolyte and blood pressure homeostasis, but also in airway surface liquid homeostasis, which is important for proper clearance of mucus. Controls the reabsorption of sodium in kidney, colon, lung and sweat glands. Also plays a role in taste perception.
Type: Primary
Antigen: SCNN1B Ser633
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse