Anti-DPH1 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-5714R-CY5.5
BOSSBS-5714R-CY5.5EA 900 EUR
BOSSBS-5714R-CY5.5
Anti-DPH1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
OVCA1 is required for the first step in the synthesis of diphthamide, a unique posttranslationally modified histidine found only in translation elongation factor-2 (EEF2; MIM 130610). This modification is conserved from archaebacteria to humans and serves as the target for ADP-ribosylation and inactivation of EEF2 by diphtheria toxin (DT) and Pseudomonas exotoxin A. OVCA1 is one of several enzymes involved in synthesis of diphthamide in EEF2.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DPH1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR