Anti-RASL10A Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-5722R-A488
BOSSBS-5722R-A488EA 893 EUR
BOSSBS-5722R-A488
Anti-RASL10A Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
Ras proteins are members of a superfamily of related small GTPases. RRP22 can be posttranslationally modified by farnesyl. RRP22 inhibits cell growth and promotes caspase-independent cell death. The specific function of RASL10A is not yet known.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RASL10A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR