Anti-RASL10A Rabbit Polyclonal Antibody (Cy5.5®)
Lieferant: Bioss
BS-5722R-CY5.5
BOSSBS-5722R-CY5.5EA 900 EUR
BOSSBS-5722R-CY5.5
Anti-RASL10A Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Ras proteins are members of a superfamily of related small GTPases. RRP22 can be posttranslationally modified by farnesyl. RRP22 inhibits cell growth and promotes caspase-independent cell death. The specific function of RASL10A is not yet known.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RASL10A
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat