Anti-PARD6A Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss
BS-5733R-FITC
BOSSBS-5733R-FITCEA 1350 EUR
BOSSBS-5733R-FITC
Anti-PARD6A Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Adapter protein involved in asymmetrical cell division and cell polarization processes. Probably involved in the formation of epithelial tight junctions. Association with PARD3 may prevent the interaction of PARD3 with F11R/JAM1, thereby preventing tight junction assembly. The PARD6-PARD3 complex links GTP-bound Rho small GTPases to atypical protein kinase C proteins.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PARD6A
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR