Anti-DCK Rabbit Polyclonal Antibody

Lieferant: Bioss
BS-5749R
BOSSBS-5749REA 725 EUR
BOSSBS-5749R
Anti-DCK Rabbit Polyclonal Antibody
Antikörper
Deoxycytidine kinase is responsible for the phosphorylation of several deoxyribonucleosides and their analogs. Deficiency of this enzyme activity is associated with resistance to antiviral and anticancer chemotherapeutic agents, whereas increased enzyme activity is associated with increased activation of these compounds to cytotoxic nucleoside triphosphate derivatives. It is the rate limiting enzyme in the activation of many important anticancer and retroviral drugs and its activity is often decreased in cells that are resistant to cytosine arabinoside.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: DCK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR