Anti-ABCC10 Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-5761R-CY5
BOSSBS-5761R-CY5EA 900 EUR
BOSSBS-5761R-CY5
Anti-ABCC10 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, and White). This ABC full-transporter is a member of the MRP subfamily which is involved in multi ATP-dependent transporter probably involved in cellular detoxification through lipophilic anion extrusion.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ABCC10
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat