Anti-ABCC10 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-5761R-CY7
BOSSBS-5761R-CY7EA 900 EUR
BOSSBS-5761R-CY7
Anti-ABCC10 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, and White). This ABC full-transporter is a member of the MRP subfamily which is involved in multi ATP-dependent transporter probably involved in cellular detoxification through lipophilic anion extrusion.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ABCC10
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR