Anti-LANCL2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Lieferant: Bioss
BS-5765R-A680
BOSSBS-5765R-A680EA 890 EUR
BOSSBS-5765R-A680
Anti-LANCL2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antikörper
Necessary for abscisic acid (ABA) binding on the cell membrane and activation of the ABA signaling pathway in granulocytes.
Type: Primary
Antigen: LANCL2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 331-430/460
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen