Anti-Activin A Receptor Type IC Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-5777R-A750
BOSSBS-5777R-A750EA 890 EUR
BOSSBS-5777R-A750
Anti-Activin A Receptor Type IC Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Serine/threonine protein kinase which forms a receptor complex on ligand binding. The receptor complex consisting of 2 type II and 2 type I transmembrane serine/threonine kinases. Type II receptors phosphorylate and activate type I receptors which autophosphorylate, then bind and activate SMAD transcriptional regulators, SMAD2 and SMAD3. Receptor for activin AB, activin B and NODAL. Plays a role in cell differentiation, growth arrest and apoptosis.

Type: Primary
Antigen: Activin A Receptor Type IC
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 21-120/493
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR