Anti-ACVR1C Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-5777R-CY5.5
BOSSBS-5777R-CY5.5EA 900 EUR
BOSSBS-5777R-CY5.5
Anti-ACVR1C Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
Serine/threonine protein kinase which forms a receptor complex on ligand binding. The receptor complex consisting of 2 type II and 2 type I transmembrane serine/threonine kinases. Type II receptors phosphorylate and activate type I receptors which autophosphorylate, then bind and activate SMAD transcriptional regulators, SMAD2 and SMAD3. Receptor for activin AB, activin B and NODAL. Plays a role in cell differentiation, growth arrest and apoptosis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ACVR1C
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
More Product Information
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR