Anti-ELMO1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Lieferant: Bioss
BS-5807R-A647
BOSSBS-5807R-A647EA 893 EUR
BOSSBS-5807R-A647
Anti-ELMO1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antikörper
ELMO1 is involved in cytoskeletal rearrangements required for phagocytosis of apoptotic cells and cell motility. ELMO1 acts in association with DOCK1 and CRK and was initially proposed to be required in complex with DOCK1 to activate Rac Rho small GTPases. It is belived to enhance the guanine nucleotide exchange factor (GEF) activity of DOCK1.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ELMO1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR