Anti-ELMO1 Rabbit Polyclonal Antibody (Cy5®)
Lieferant: Bioss
BS-5807R-CY5
BOSSBS-5807R-CY5EA 900 EUR
BOSSBS-5807R-CY5
Anti-ELMO1 Rabbit Polyclonal Antibody (Cy5®)
Antikörper
ELMO1 is involved in cytoskeletal rearrangements required for phagocytosis of apoptotic cells and cell motility. ELMO1 acts in association with DOCK1 and CRK and was initially proposed to be required in complex with DOCK1 to activate Rac Rho small GTPases. It is belived to enhance the guanine nucleotide exchange factor (GEF) activity of DOCK1.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ELMO1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat