Anti-CDO1 Rabbit Polyclonal Antibody (Cy5.5®)

Lieferant: Bioss
BS-5808R-CY5.5
BOSSBS-5808R-CY5.5EA 900 EUR
BOSSBS-5808R-CY5.5
Anti-CDO1 Rabbit Polyclonal Antibody (Cy5.5®)
Antikörper
CDO1 (cysteine dioxygenase, type I) is a 200 amino acid protein that belongs to the cysteine dioxygenase family and is involved in organosulfur biosynthesis. Existing as a monomer and expressed at high levels in liver and placenta and at lower levels in brain, pancreas and heart, CDO1 functions as a dioxygenase that uses iron and zinc as cofactors to catalyze the conversion of L-cysteine and oxygen to 3-sulfinoalanine. Via its catalytic activity, CDO1 is involved in pyruvate-, sulfate- and taurine-related metabolic pathways and is a crucial regulator of cysteine concentrations within the cell. Human CDO1 shares 94% amino acid identity with its rat counterpart, suggesting a conserved role between species. The gene encoding CDO1 maps to human chromosome 5, which contains 181 million base pairs and comprises nearly 6% of the human genome. Deletion of the p arm of chromosome 5 leads to Cri du chat syndrome, while deletion of the q arm or of chromosome 5 altogether is common in therapy-related acute myelogenous leukemias and myelodysplastic syndrome.PathwayOrganosulfur biosynthesis; taurine biosynthesis; hypotaurine from L-cysteine: step 1/2.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CDO1
Clonality: Polyclonal
Clone: 0
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR