Anti-MGAT5 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Lieferant: Bioss
BS-5841R-A555
BOSSBS-5841R-A555EA 893 EUR
BOSSBS-5841R-A555
Anti-MGAT5 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
MGAT5 is a mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase, a glycosyltransferase involved in the synthesis of protein-bound and lipid-bound oligosaccharides. Alterations of the oligosaccharides on cell surface glycoproteins cause significant changes in the adhesive or migratory behavior of a cell. Increase in the encoded protein's activity may correlate with the progression of invasive malignancies.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MGAT5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat