Anti-MGAT5 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-5841R-CY7
BOSSBS-5841R-CY7EA 900 EUR
BOSSBS-5841R-CY7
Anti-MGAT5 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
MGAT5 is a mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase, a glycosyltransferase involved in the synthesis of protein-bound and lipid-bound oligosaccharides. Alterations of the oligosaccharides on cell surface glycoproteins cause significant changes in the adhesive or migratory behavior of a cell. Increase in the encoded protein's activity may correlate with the progression of invasive malignancies.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MGAT5
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR