Anti-MUC7 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Lieferant: Bioss
BS-5879R-A350
BOSSBS-5879R-A350EA 893 EUR
BOSSBS-5879R-A350
Anti-MUC7 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antikörper
May function in a protective capacity by promoting the clearance of bacteria in the oral cavity and aiding in mastication, speech, and swallowing. Binds P.aeruginosa pili.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MUC7
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR