Anti-ADORA2B Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Bioss
BS-5900R-HRP
BOSSBS-5900R-HRPEA 900 EUR
BOSSBS-5900R-HRP
Anti-ADORA2B Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Receptor for adenosine. The activity of this receptor is mediated by G proteins which activate adenylyl cyclase.

Recommended Dilutions: Western Blot: 1:100-1000

Type: Primary
Antigen: ADORA2B
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR