Anti-PPIA Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Lieferant: Bioss
BS-5912R-A488
BOSSBS-5912R-A488EA 915 EUR
BOSSBS-5912R-A488
Anti-PPIA Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antikörper
PPIases accelerate the folding of proteins. It catalyzes the cis-trans isomerization of proline imidic peptide bonds in oligopeptides.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PPIA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR