Anti-PPIA Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Lieferant: Bioss
BS-5912R-A555
BOSSBS-5912R-A555EA 915 EUR
BOSSBS-5912R-A555
Anti-PPIA Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antikörper
PPIases accelerate the folding of proteins. It catalyzes the cis-trans isomerization of proline imidic peptide bonds in oligopeptides.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PPIA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat