Anti-CYPA Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Lieferant: Bioss
BS-5912R-A750
BOSSBS-5912R-A750EA 913 EUR
BOSSBS-5912R-A750
Anti-CYPA Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
PPIases accelerate the folding of proteins. It catalyzes the cis-trans isomerisation of proline imidic peptide bonds in oligopeptides.

Type: Primary
Antigen: CYPA
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-165/165
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR