Anti-SERINC3 Rabbit Polyclonal Antibody (Cy7®)

Lieferant: Bioss
BS-5942R-CY7
BOSSBS-5942R-CY7EA 900 EUR
BOSSBS-5942R-CY7
Anti-SERINC3 Rabbit Polyclonal Antibody (Cy7®)
Antikörper
TDE1, tumor differentially expressed protein 1, is a multi-pass membrane protein also known as serine incorporator 3. Also found on the membranes of the Golgi apparatus within cells, TDE1 is highly expressed in neuronal populations but is also found in thymus, kidney, liver and testis. Expression levels of TDE1 in tumors can be as much as tenfold the amount found in normal tissue of the same type. This increased expression implicates TDE1 as being involved in the cellular transformation from normal to malignant tissue. It is believed TDE1 contributes to oncogenesis by partially protecting cells from serum starvation and etoposide-induced apoptosis. The mechanism through which TDE1 protects cells is poorly understood, but may involve aberrant methylation of TDE1 complexes.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SERINC3
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR