Anti-LIMS1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Lieferant: Bioss
BS-5951R-A750
BOSSBS-5951R-A750EA 890 EUR
BOSSBS-5951R-A750
Anti-LIMS1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antikörper
Effector of integrin and growth factor signaling, coupling surface receptors to downstream signaling molecules involved in the regulation of cell survival, cell proliferation and cell differentiation. Focal adhesion protein part of the complex ILK-PINCH. This complex is considered to be one of the convergence points of integrin- and growth factor-signaling pathway.
Type: Primary
Antigen: LIMS1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/325
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen