Anti-N-Myc Rabbit Polyclonal Antibody (Cy5®)

Lieferant: Bioss
BS-5980R-CY5
BOSSBS-5980R-CY5EA 1350 EUR
BOSSBS-5980R-CY5
Anti-N-Myc Rabbit Polyclonal Antibody (Cy5®)
Antikörper
This gene is a member of the MYC family and encodes a protein with a basic helix-loop-helix (bHLH) domain. This protein is located in the nucleus and must dimerize with another bHLH protein in order to bind DNA. Amplification of this gene is associated with a variety of tumors, most notably neuroblastomas. [provided by RefSeq, Jul 2008].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: n-Myc
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR